3-Amino-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A251699-100MG A251699-10G A251699-250MG A251699-1G A251699-5G
103812-000EA 23 USD
103812-000 103851-840 77031-532 77031-534 77031-536 77091-228 77091-230
3-Amino-5-(trifluoromethyl)benzonitrile 98%
3-Amino-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C8H5F3N2
MW: 186.1339 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09955455
CAS Number: 49674-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR