3-Amino-4-pyridazinecarboxylic acid 95%

Supplier: Ambeed
Warning

A118437-100MG A118437-250MG A118437-5G A118437-25G
103799-058EA 64 USD
103799-058 77009-282 77009-284 77311-730 77683-066
3-Amino-4-pyridazinecarboxylic acid 95%
3-Amino-4-pyridazinecarboxylic acid

Precaución: For research use only.

Formula: C5H5N3O2
MW: 139.1100 g/mol
Boiling Pt: 470.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01318258
CAS Number: 21141-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR