3-Amino-4-methylpyridine 98%

Supplier: Ambeed
Warning

A549827-5G A549827-100G A549827-25G
103839-816EA 5 USD
103839-816 76843-886 77279-978 77307-792
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MW: 108.14
Boiling Pt: 261.3 °C (760 mm Hg)
Melting Pt: 102…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00128871
CAS Number: 3430-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR