3-Amino-4-methylphenol 98%

Supplier: Ambeed
Warning

A283333-1G A283333-25G A283333-5G A283333-10G A283333-250MG A283333-100G
103813-610EA 7 USD
103813-610 103853-560 76837-264 76837-266 76837-268 77002-214 77002-216 77550-738
3-Amino-4-methylphenol 98%
3-Amino-4-methylphenol

Precaución: for Research Use Only

Formula: C₇H₉NO
Boiling Pt: 259 °C (760 mm Hg)
Melting Pt: 159 °C
Flash Pt: 119.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00075083
CAS Number: 2836-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR