3-Amino-4-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A233024-100G A233024-10G A233024-25G A233024-5G A233024-1G
103810-868EA 9 USD
103810-868 103850-626 76815-198 76815-200 77089-574 77089-576 77303-952
3-Amino-4-methylbenzonitrile 98%
3-Amino-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Flash Pt: 142.987 °C
Storage Temperature: Ambient
MDL Number: MFCD04039781
CAS Number: 60710-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR