3-Amino-4-iodopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodo-3-pyridylamine, 4-Iodopyridin-3-amine, 4-Iodo-pyridin-3-ylamine

A101078-10G A101078-250MG A101078-25G A101078-1G A101078-5G
103795-794EA 23 USD
103795-794 103821-484 77008-304 77008-306 77008-308 77282-652 77282-654
3-Amino-4-iodopyridine 95%
3-Amino-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
Melting Pt: 65…70 °C
Storage Temperature: Ambient
MDL Number: MFCD04971334
CAS Number: 105752-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR