3-Amino-4-hydroxypyridine 95%

Supplier: Ambeed
Warning

A218309-1G A218309-25G A218309-5G A218309-100G A218309-10G
103818-704EA 9 USD
103818-704 103849-578 77088-098 77088-100 77088-102 77288-140 77288-142
3-Amino-4-hydroxypyridine 95%
3-Amino-4-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₆N₂O
Boiling Pt: 253.6 °C (760 mm Hg)
Melting Pt: 145…149 °C
Storage Temperature: Ambient
MDL Number: MFCD04114230
CAS Number: 6320-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR