3-Amino-4-hydroxybenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A135434-100G
A135434-10G
A135434-1G
A135434-25G
A135434-5G
A135434-500G
103802-154EA
7
USD
103802-154
76767-440
76767-442
77314-976
77314-978
77314-980
77596-864
3-Amino-4-hydroxybenzoic acid 97%
3-Amino-4-hydroxybenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₃ Boiling Pt: 388.8 °C (760 mm Hg) Flash Pt: 189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007697 CAS Number: 1571-72-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...