3-Amino-4-fluorobenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A152045-100G
A152045-10G
A152045-1G
A152045-25G
A152045-5G
103804-344EA
11
USD
103804-344
103842-498
76871-638
76871-640
77318-100
77318-102
77318-104
3-Amino-4-fluorobenzonitrile 98%
3-Amino-4-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787428 CAS Number: 859855-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...