3-Amino-4-chloropyridine 98%

Supplier: Ambeed
Warning

A102090-100G A102090-10G A102090-1G A102090-25G A102090-5G
103796-292EA 9 USD
103796-292 103821-986 77282-900 77282-902 77282-904 77282-906 77308-874
3-Amino-4-chloropyridine 98%
3-Amino-4-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
Boiling Pt: 247.6 °C (760 mm Hg)
Flash Pt: 103.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235124
CAS Number: 20511-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR