3-Amino-4-chlorophenol 98%
Supplier: Ambeed
|
Warning
|
A217744-1G
A217744-25G
A217744-5G
103818-644EA
13
USD
103818-644
103849-374
77088-002
77088-004
77088-006
3-Amino-4-chlorophenol 98%
3-Amino-4-chlorophenol
Precaución: for Research Use Only
Formula:
C₆H₆ClNO MW: 143.57 g/mol Boiling Pt: 282.8 °C (760 mm Hg) Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427127 CAS Number: 16026-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...