3-Amino-4-bromopyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-3-pyridylamine, 4-Bromopyridin-3-amine
A113373-10G
A113373-250MG
A113373-25G
A113373-1G
A113373-5G
103798-222EA
19
USD
103798-222
103824-064
76995-354
76995-356
76995-358
77310-882
77310-884
3-Amino-4-bromopyridine 97%
3-Amino-4-bromopyridine
Precaución: for Research Use Only
Formula:
C₅H₅BrN₂ Boiling Pt: 287.6 °C (760 mm Hg) Flash Pt: 127.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD04112575 CAS Number: 239137-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...