3-Amino-4-bromophenol 98%

Supplier: Ambeed
Warning

A608264-1G A608264-5G A608264-10G A608264-250MG A608264-100G
103840-152EA 45 USD
103840-152 76844-316 76844-318 76980-544 76980-546 77673-648
3-Amino-4-bromophenol 98%
3-Amino-4-bromophenol

Precaución: For research use only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Boiling Pt: 300.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09260886
CAS Number: 100367-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR