3-Amino-4-bromobenzonitrile 98%

Supplier: Ambeed
Warning

A379933-1G A379933-25G A379933-5G A379933-100MG A379933-10G A379933-250MG
103826-656EA 53 USD
103826-656 103837-692 76841-012 76841-014 76841-016 77016-056 77016-058 77016-060
3-Amino-4-bromobenzonitrile 98%
3-Amino-4-bromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Boiling Pt: 315.2±22.0 °C (760 mm Hg)
Melting Pt: 99 °C
Flash Pt: 144.4 °C
Storage Temperature: Ambient
MDL Number: MFCD11036380
CAS Number: 72635-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR