3-Amino-4-bromo-6-chloropyridazine 97%
Supplier: Ambeed
|
Warning
|
A118109-100G
A118109-10G
A118109-250MG
A118109-25G
A118109-500G
A118109-1G
A118109-5G
103798-994EA
7
USD
103798-994
103824-886
76781-524
76781-526
76781-528
76781-530
76781-532
77311-674
77311-676
3-Amino-4-bromo-6-chloropyridazine 97%
3-Amino-4-bromo-6-chloropyridazine
Precaución: for Research Use Only
Formula:
C₄H₃BrClN₃ Flash Pt: 186.865 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11520891 CAS Number: 446273-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...