3-Amino-4-(piperidin-1-yl)benzoic acid 95%
Supplier: Ambeed
|
Warning
|
A376760-1G
A376760-250MG
103826-592EA
106
USD
103826-592
103837-618
76840-922
77278-496
3-Amino-4-(piperidin-1-yl)benzoic acid 95%
3-Amino-4-(piperidin-1-yl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₆N₂O₂ MW: 220.27 g/mol Boiling Pt: 443.9 °C (760 mm Hg) Flash Pt: 222.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD02225752 CAS Number: 26586-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...