3-Amino-4-(methylamino)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A267557-1G
A267557-100MG
A267557-250MG
A267557-50MG
A267557-5G
103812-862EA
68
USD
103812-862
103852-762
77092-510
77276-716
77276-718
77276-720
77304-980
3-Amino-4-(methylamino)benzoic acid 97%
3-Amino-4-(methylamino)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Boiling Pt: 386 °C (760 mm Hg) Flash Pt: 187.251 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06208249 CAS Number: 66315-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...