3-Amino-3-methylbutanoic acid 98%

Supplier: Ambeed
Warning

A317374-1G A317374-5G A317374-100G A317374-10G
103825-132EA 8 USD
103825-132 103836-064 76838-912 76838-914 76989-068 77885-846
3-Amino-3-methylbutanoic acid 98%
3-Amino-3-methylbutanoic acid

Precaución: for Research Use Only

Formula: C5H11NO2
Boiling Pt: 435.9 °C (760 mm Hg)
Flash Pt: 87.259 °C
Storage Temperature: Ambient
MDL Number: MFCD06245495
CAS Number: 625-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR