3-Amino-3-methylbutan-1-ol 97%

Supplier: Ambeed
Warning

A305490-5G A305490-100MG A305490-10G A305490-250MG A305490-25G
103814-434EA 39 USD
103814-434 103835-600 76838-300 77277-402 77277-404 77277-406 77305-912
3-Amino-3-methylbutan-1-ol 97%
3-Amino-3-methylbutan-1-ol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 179.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09032902
CAS Number: 42514-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR