3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid 97%

Supplier: Ambeed
Warning

A119868-250MG A119868-1G
103799-850EA 26 USD
103799-850 77271-428 77311-958
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid 97%
3-Amino-3-[3-(trifluoromethyl)phenyl]propanoic acid

Precaución: for Research Use Only

Formula: C10H10F3NO2
MW: 233.1900 g/mol
Boiling Pt: 295.2 °C (760 mm Hg)
Melting Pt: 210…212 °C
Flash Pt: 132.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02653329
CAS Number: 143438-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR