3-Amino-3-(4-bromophenyl)propionic acid 95%
Supplier: Ambeed
|
Warning
|
A145050-100G
A145050-10G
A145050-250MG
A145050-1G
103803-488EA
7
USD
103803-488
103841-570
76811-936
76811-938
76811-940
77316-842
3-Amino-3-(4-bromophenyl)propionic acid 95%
3-Amino-3-(4-bromophenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀BrNO₂ MW: 244.0900 g/mol Boiling Pt: 378.1 °C (760 mm Hg) Melting Pt: 235…236 °C Flash Pt: 182.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00187112 CAS Number: 39773-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...