2-Fluoro-m-toluidine 97%

Supplier: Ambeed
Warning

A166467-10G A166467-1G A166467-25G A166467-5G
103806-072EA 9 USD
103806-072 103844-334 76768-836 77300-842 77320-584 77320-586
2-Fluoro-m-toluidine 97%
2-Fluoro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈FN
Boiling Pt: 187.6 °C (760 mm Hg)
Flash Pt: 80.338 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06410915
CAS Number: 1978-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR