2-Fluoro-3-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, 3-Amino-2-fluorobenzotrifluoride, α,α,α,2-Tetrafluoro-m-toluidine, 3-Amino-α,α,α,2-tetrafluorotoluene

A212054-1G A212054-25G A212054-5G A212054-100G A212054-10G
103818-162EA 10 USD
103818-162 103848-858 77087-302 77087-304 77087-306 77139-618 77139-620
2-Fluoro-3-(trifluoromethyl)aniline 97%
2-Fluoro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅F₄N
MW: 179.12
Boiling Pt: 189
Density: 1.39
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061241
CAS Number: 123973-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR