3-Amino-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A105329-250MG A105329-1G
103796-870EA 34 USD
103796-870 103822-600 77135-678 77309-470
3-Amino-2-fluorobenzoic acid 98%
3-Amino-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C7H6FNO2
MW: 155.1300 g/mol
Boiling Pt: 325.9 °C (760 mm Hg)
Flash Pt: 150.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06797827
CAS Number: 914223-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR