2-Chloro-6-methoxypyridin-3-amine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-2-chloro-6-methoxypyridine

A147101-10G A147101-25G A147101-5G
103803-736EA 12 USD
103803-736 103841-832 77259-084 77317-188 77317-190
2-Chloro-6-methoxypyridin-3-amine 98%
2-Chloro-6-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.59 g/mol
Boiling Pt: 366.6 °C (760 mm Hg)
Melting Pt: 46…50 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258809
CAS Number: 34392-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR