3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A111221-10G A111221-1G A111221-5G
103797-808EA 35 USD
103797-808 103823-622 77148-706 77310-426 77310-428
3-Amino-2-chloro-6-(trifluoromethyl)pyridine 98%
3-Amino-2-chloro-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C6H4ClF3N2
MW: 196.5600 g/mol
Boiling Pt: 231.8 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862661
CAS Number: 117519-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR