2-Bromo-3-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A233294-1G A233294-25G A233294-5G A233294-100G A233294-10G
103810-880EA 7 USD
103810-880 103850-640 77089-588 77089-590 77089-592 77261-588 77261-590
2-Bromo-3-(trifluoromethyl)aniline 98%
2-Bromo-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.02
Boiling Pt: 228.9 °C (760 mm Hg)
Flash Pt: 92.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01656033
CAS Number: 58458-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR