3-Amino-2-bromo-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A364683-1G A364683-100G A364683-100MG A364683-10G A364683-250MG A364683-25G A364683-5G
103826-366EA 9 USD
103826-366 103837-386 76840-616 77115-334 77115-336 77115-338 77115-340 77115-342 77306-760
3-Amino-2-bromo-5-fluoropyridine 98%
3-Amino-2-bromo-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
Boiling Pt: 273.6 °C (760 mm Hg)
Flash Pt: 119.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05662413
CAS Number: 884495-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR