3-Amino-2,6-dichloropyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichloro-3-pyridinamine, 2,6-Dichloropyridin-3-amine, 2,6-Dichloro-3-pyridylamine
A480304-1G
A480304-5G
A480304-100G
A480304-10G
103828-118EA
11
USD
103828-118
103839-194
76843-134
76843-136
76991-000
76991-002
3-Amino-2,6-dichloropyridine 97%
3-Amino-2,6-dichloropyridine
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Boiling Pt: 305.3 °C (760 mm Hg) Melting Pt: 122 °C Flash Pt: 138.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023417 CAS Number: 62476-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...