3-Amino-2,6-dichloroisonicotinic acid 95%
Supplier: Ambeed
|
Warning
|
A194240-250MG
A194240-1G
A194240-10G
103816-636EA
51
USD
103816-636
103847-248
76986-226
77302-402
77683-519
3-Amino-2,6-dichloroisonicotinic acid 95%
3-Amino-2,6-dichloroisonicotinic acid
Precaución: For research use only.
Formula:
C₆H₄Cl₂N₂O₂ MW: 207.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24445281 CAS Number: 58484-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...