3-Amino-2,4-dichloropyridine 98%

Supplier: Ambeed
Warning

A112576-100G A112576-10G A112576-250MG A112576-25G A112576-1G A112576-5G
103798-042EA 19 USD
103798-042 103823-880 77245-774 77245-776 77245-778 77245-780 77310-688 77310-690
3-Amino-2,4-dichloropyridine 98%
3-Amino-2,4-dichloropyridine

Precaución: for Research Use Only

Formula: C5H4Cl2N2
MW: 163.01 g/mol
Boiling Pt: 264.9 °C (760 mm Hg)
Flash Pt: 114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750301
CAS Number: 173772-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR