3-Amino-2,2-difluoropropan-1-ol 95%

Supplier: Ambeed
Danger

Synonyms: 2,2-Difluoro-3-hydroxypropan-1-amine

A226725-5G A226725-250MG
103819-266EA 50 USD
103819-266 103850-170 77088-918 77152-818
3-Amino-2,2-difluoropropan-1-ol 95%
3-Amino-2,2-difluoropropan-1-ol

Precaución: for Research Use Only

Formula: C₃H₇F₂NO
MW: 111.091 g/mol
Boiling Pt: 216.2±35.0 °C (760 mm Hg)
Flash Pt: 84.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19207959
CAS Number: 155310-11-5
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR