3-Acetylthiophene 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-thienyl ketone, 1-(3-Thienyl)-1-ethanone

A275565-25G A275565-5G A275565-100G A275565-10G A275565-1G
103813-264EA 5 USD
103813-264 103853-188 77093-042 77093-044 77153-710 77153-712 77305-200
3-Acetylthiophene 98%
3-Acetylthiophene

Precaución: for Research Use Only

Formula: C₆H₆OS
MW: 126.18 g/mol
Boiling Pt: 190.5 °C (760 mm Hg)
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00005468
CAS Number: 1468-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR