3-Acetylphenylboronic acid 97%

Supplier: Ambeed
Warning

A234658-25G A234658-5G A234658-100G A234658-10G A234658-1G
103810-966EA 6 USD
103810-966 103850-722 77089-714 77089-716 77249-628 77249-630 77303-990
3-Acetylphenylboronic acid 97%
3-Acetylphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
Boiling Pt: 364 °C (760 mm Hg)
Melting Pt: 204…208 °C
Flash Pt: 173.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01074678
CAS Number: 204841-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR