3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene 97%

Supplier: Ambeed
Warning

A294593-100G A294593-10G A294593-1G A294593-25G A294593-5G
103814-012EA 13 USD
103814-012 103835-152 76827-758 76827-760 77305-652 77305-654 77305-656
3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene 97%
3-Acetyl-2-(aminosulfonyl)-5-chlorothiophene

Precaución: for Research Use Only

Formula: C₆H₆ClNO₃S₂
Boiling Pt: 453 °C (760 mm Hg)
Flash Pt: 227.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09033309
CAS Number: 160982-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR