3-Acetyl-5-bromopyridine 97%

Supplier: Ambeed
Warning

A123564-100G A123564-10G A123564-250MG A123564-25G A123564-1G A123564-5G
103800-416EA 6 USD
103800-416 77161-466 77161-468 77161-470 77161-472 77312-534 77312-536
3-Acetyl-5-bromopyridine 97%
3-Acetyl-5-bromopyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO
MW: 200.03 g/mol
Boiling Pt: 279.3 °C (760 mm Hg)
Melting Pt: 77…79 °C
Flash Pt: 122.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03086033
CAS Number: 38940-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR