1-(2,6-Dichloropyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

Synonyms: 3-Acetyl-2,6-dichloropyridine

A125388-100MG A125388-250MG A125388-1G A125388-5G
103800-672EA 72 USD
103800-672 77026-750 77026-752 77298-046 77298-048
1-(2,6-Dichloropyridin-3-yl)ethanone 95%
1-(2,6-Dichloropyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO
MW: 190.0267 g/mol
Boiling Pt: 286.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11847148
CAS Number: 412018-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR