3-Acetyl-2,5-dichlorothiophene 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Dichloro-3-thienyl)-1-ethanone

A720498-25G A720498-5G A720498-100G
103840-738EA 11 USD
103840-738 76844-952 76844-954 77686-718
3-Acetyl-2,5-dichlorothiophene 98%
3-Acetyl-2,5-dichlorothiophene

Precaución: for Research Use Only

Formula: C₆H₄Cl₂OS
MW: 195.07 g/mol
Boiling Pt: 233.9 °C (760 mm Hg)
Melting Pt: 37…39 °C at N/A mmHg
Flash Pt: 95.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014522
CAS Number: 36157-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR