(3-Acetamidophenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Acetamidobenzeneboronic acid, 3-Acetamidophenylboronic acid

A171990-1G A171990-25G A171990-5G A171990-100G A171990-10G A171990-250MG
103806-686EA 9 USD
103806-686 103844-968 77082-144 77082-146 77082-148 77191-444 77191-446 77191-448
(3-Acetamidophenyl)boronic acid 95%
(3-Acetamidophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₃
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00236013
CAS Number: 78887-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR