3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulfonic Acid Hydrate 98%

Supplier: Ambeed
Danger

Synonyms: 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulphonic acid Hydrate

A938790-10G A938790-25G A938790-5G
103841-424EA 9 USD
103841-424 76820-996 76845-690 76845-692
3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulfonic Acid Hydrate 98%
3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulfonic Acid Hydrate

Precaución: for Research Use Only

Formula: C₉H₂₂N₂O₆S
MW: 286.34 g/mol
Melting Pt: 158…160 °C at N/A mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00150550
CAS Number: 865856-46-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR