3-[2-(Trifluoromethyl)phenyl]propionic acid 98%

Supplier: Ambeed
Warning

A102294-10G A102294-5G A102294-25G
103796-354EA 8 USD
103796-354 103822-046 76970-218 76970-220 77308-928 77308-930 77880-832
3-[2-(Trifluoromethyl)phenyl]propionic acid 98%
3-[2-(Trifluoromethyl)phenyl]propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
Boiling Pt: 266.9 °C (760 mm Hg)
Melting Pt: 84…88 °C
Flash Pt: 115.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06823965
CAS Number: 94022-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR