Boc-(3-aminomethylphenyl)acetic acid 96%

Supplier: Ambeed
Warning

Synonyms: 3-[(Boc-amino)methyl]phenylacetic Acid

A135173-100MG A135173-250MG A135173-5G
103802-102EA 86 USD
103802-102 76767-422 76767-424 77298-756
Boc-(3-aminomethylphenyl)acetic acid 96%
Boc-(3-aminomethylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Boiling Pt: 435.2 °C (760 mm Hg)
Flash Pt: 217 °C
Storage Temperature: Ambient
MDL Number: MFCD06656429
CAS Number: 71420-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR