3,8-Dibromo-1,10-phenanthroline 97%

Supplier: Ambeed
Warning

A174915-1G A174915-10G A174915-250MG
103806-988EA 66 USD
103806-988 103845-288 77082-600 77273-836 77273-838
3,8-Dibromo-1,10-phenanthroline 97%
3,8-Dibromo-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C12H6Br2N2
Flash Pt: 213.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09909860
CAS Number: 100125-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR