3,6-Di-tert-butylcarbazole 98%

Supplier: Ambeed
Warning

A164322-100G A164322-10G A164322-500G A164322-1G A164322-25G A164322-5G
103805-844EA 5 USD
103805-844 103844-104 76812-940 76812-942 76812-944 77300-708 77300-710 77300-712
3,6-Di-tert-butylcarbazole 98%
3,6-Di-tert-butylcarbazole

Precaución: for Research Use Only

Formula: C₂₀H₂₅N
Boiling Pt: 424.2±14.0 °C (760 mm Hg)
Melting Pt: 228…233 °C
Flash Pt: 181.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03425849
CAS Number: 37500-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR