3,6-Diphenylcarbazole 98%

Supplier: Ambeed
Warning

Synonyms: 3,6-Diphenyl-9H-carbazole

A113167-100G A113167-10G A113167-25G A113167-1G A113167-5G
103798-172EA 6 USD
103798-172 103824-014 77271-024 77271-026 77271-028 77310-842 77310-844
3,6-Diphenylcarbazole 98%
3,6-Diphenylcarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₇N
MW: 319.41 g/mol
Boiling Pt: 575.4 °C (760 mm Hg)
Melting Pt: 186 °C
Flash Pt: 256 °C
Storage Temperature: Ambient
MDL Number: MFCD00222619
CAS Number: 56525-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR