2,2'-[ethylenebis(oxy)]bisacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dioxaoctanedioic acid

A443293-250MG A443293-25G
103827-694EA 30 USD
103827-694 103838-764 77279-144 77551-336
2,2'-[ethylenebis(oxy)]bisacetic acid 97%
2,2'-[ethylenebis(oxy)]bisacetic acid

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 429.2 °C (760 mm Hg)
Melting Pt: 68…71 °C
Flash Pt: 182.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00054502
CAS Number: 23243-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR