3,4-Epoxytetrahydrofuran 97%
Supplier: Ambeed
|
Danger
|
A137229-10G
A137229-1G
A137229-25G
103802-438EA
5
USD
103802-438
77285-588
77315-382
77315-384
3,4-Epoxytetrahydrofuran 97%
3,4-Epoxytetrahydrofuran
Precaución: for Research Use Only
Formula:
C₄H₆O₂ Boiling Pt: 128 °C (760 mm Hg) Flash Pt: 31.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800639 CAS Number: 285-69-8 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...