3,6-Dimethylpyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A170440-250MG A170440-100MG
103806-500EA 140 USD
103806-500 76824-472 77885-338
3,6-Dimethylpyrazine-2-carboxylic acid 97%
3,6-Dimethylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13174793
CAS Number: 2435-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR