3,6-Dimethylpyrazin-2-amine 97%

Supplier: Ambeed
Warning

A250613-100MG A250613-250MG A250613-1G
103811-936EA 53 USD
103811-936 103851-774 76874-620 76874-622 77091-128
3,6-Dimethylpyrazin-2-amine 97%
3,6-Dimethylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.156 g/mol
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD18917551
CAS Number: 13134-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR