3,6-Dimethyl-2-pyridinamine 98%

Supplier: Ambeed
Warning

A128370-250MG A128370-1G A128370-5G
103801-114EA 55 USD
103801-114 76870-626 77298-296 77298-298
3,6-Dimethyl-2-pyridinamine 98%
3,6-Dimethyl-2-pyridinamine

Precaución: for Research Use Only

Formula: C7H10N2
MW: 122.1700 g/mol
Boiling Pt: 223 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD06637718
CAS Number: 823-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR